Assay Details
Target Gene Details
Entrez Gene ID: | 12892 |
Gene Name: | coproporphyrinogen oxidase |
Gene Aliases: |
CPX, Cpo, HCP, cac, nct |
Location: |
Chr.16:58670208-58680391 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpox | NM_007757.2 | NP_031783.2 | ||
| AK089471.1 | ||||
| AK166512.1 | ||||
| AK187535.1 | ||||
| BC017680.1 | ||||
| D16333.1 | BAA03840.1 |
More Information
Set Membership: |
|

Genomic Map