Assay Details
Target Gene Details
Entrez Gene ID: | 110253 |
Gene Name: | TRIO and F-actin binding protein |
Gene Aliases: |
AI428493, EST478828, Tara, mKIAA1662 |
Location: |
Chr.15:78947724-79005869 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Triobp | NM_001024716.1 | NP_001019887.1 | ||
| NM_001039156.1 | NP_001034245.1 | |||
| NM_138579.4 | NP_613045.3 | |||
| AK053194.1 | BAC35308.1 | |||
| AK085512.1 | ||||
| AK129417.1 | BAC98227.2 | |||
| AK155197.1 | BAE33111.1 | |||
| AK170390.1 | BAE41764.1 | |||
| BC003984.1 | AAH03984.1 | |||
| BC042760.1 | AAH42760.1 | |||
| BC053329.1 | AAH53329.1 | |||
| CF533172.1 | ||||
| DQ228000.1 | ABB59556.1 | |||
| DQ228001.2 | ABB59557.2 | |||
| DQ278602.1 | ABB77203.1 |
More Information
Set Membership: |
|

Genomic Map