Assay Details
Target Gene Details
Entrez Gene ID: | 387609 |
Gene Name: | zinc fingers and homeoboxes 2 |
Gene Aliases: |
Afr-1, Afr1, Raf, mKIAA0854 |
Location: |
Chr.15:57694667-57839832 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zhx2 | NM_199449.2 | NP_955520.1 | ||
| AK031782.1 | BAC27546.1 | |||
| AK157631.1 | BAE34139.1 | |||
| AK214013.1 | ||||
| BC059178.1 | AAH59178.1 |
More Information
Set Membership: |
|

Genomic Map