Assay Details
Target Gene Details
Entrez Gene ID: | 56722 |
Gene Name: | LPS-induced TN factor |
Gene Aliases: |
3222402J11Rik, C85531, N4WBP3, TBX1 |
Location: |
Chr.16:10959273-10993121 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Litaf | NM_019980.2 | NP_064364.1 | ||
| AF171100.1 | AAF27312.1 | |||
| AK018578.1 | BAB31289.1 | |||
| AK076162.1 | BAC36228.1 | |||
| AK078708.1 | ||||
| AK151053.1 | BAE30070.1 | |||
| AK159533.1 | BAE35161.1 | |||
| BC018559.1 | AAH18559.1 |
More Information
Set Membership: |
|

Genomic Map