Assay Details
Target Gene Details
Entrez Gene ID: | 21976 |
Gene Name: | topoisomerase (DNA) III beta |
Gene Aliases: |
- |
Location: |
Chr.16:16870694-16892986 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Top3b | NM_011624.2 | 2 | 288 | NP_035754.1 |
| AB013603.1 | 1 | 101 | BAA34227.1 | |
| AB045324.1 | 2 | 362 | BAA97984.1 | |
| AK041159.1 | 2 | 197 | ||
| AK079765.1 | 2 | 215 | ||
| AK144709.1 | 3 | 274 | BAE26026.1 | |
| AK146475.1 | 2 | 310 | BAE27200.1 | |
| BC031723.1 | 2 | 288 | AAH31723.1 |
More Information
Set Membership: |
|

Genomic Map