Assay Details
Target Gene Details
Entrez Gene ID: | 56637 |
Gene Name: | glycogen synthase kinase 3 beta |
Gene Aliases: |
7330414F15Rik, 8430431H08Rik, C86142, GSK-3, GSK-3beta, GSK3 |
Location: |
Chr.16:38089001-38246080 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gsk3b | NM_019827.6 | NP_062801.1 | ||
AB066114.1 | BAD86827.1 | |||
AF156099.2 | AAD39258.2 | |||
AK154293.1 | BAE32494.1 | |||
AK164048.1 | 7 | 811 | ||
AK170965.1 | BAE42146.1 | |||
AK186943.1 | ||||
BC006936.1 | AAH06936.1 | |||
BC060743.1 | AAH60743.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
