Assay Details
Target Gene Details
Entrez Gene ID: | 106338 |
Gene Name: | NOL1/NOP2/Sun domain family member 3 |
Gene Aliases: |
6720484A09Rik, AU022521 |
Location: |
Chr.16:62732444-62786794 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nsun3 | AK020172.1 | |||
AK032127.1 | BAC27717.1 | |||
AK088718.1 | ||||
AK153942.1 | BAE32268.1 | |||
AK162988.1 | ||||
AK169841.1 | BAE41405.1 |
