Assay Details
Target Gene Details
Entrez Gene ID: | 67014 |
Gene Name: | myc induced nuclear antigen |
Gene Aliases: |
1810047J07Rik, 2410057H13Rik, 3830408E23Rik, AI449204 |
Location: |
Chr.16:59471769-59492461 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mina | NM_025910.3 | NP_080186.3 | ||
AB177385.1 | BAD60965.1 | |||
AK007810.1 | BAB25275.1 | |||
AK013451.1 | BAB28860.1 | |||
AK014426.1 | ||||
AK031671.1 | BAC27503.1 | |||
AK219613.1 | ||||
BC023462.1 | AAH23462.1 | |||
BC025109.1 | AAH25109.1 | |||
BC058242.1 | AAH58242.1 |
