Assay Details
Target Gene Details
Entrez Gene ID: | 224273 |
Gene Name: | beta-gamma crystallin domain containing 3 |
Gene Aliases: |
5031404N19, AU022790 |
Location: |
Chr.16:59490775-59601399 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crybg3 | NM_174848.3 | 19 | 6029 | NP_777273.2 |
| AK162506.1 | 7 | 3864 | BAE36951.1 |
Target Gene Details
Entrez Gene ID: | 67014 |
Gene Name: | myc induced nuclear antigen |
Gene Aliases: |
1810047J07Rik, 2410057H13Rik, 3830408E23Rik, AI449204 |
Location: |
Chr.16:59471769-59492461 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mina | NM_025910.3 | NP_080186.3 | ||
| AB177385.1 | BAD60965.1 | |||
| AK007810.1 | BAB25275.1 | |||
| AK013451.1 | BAB28860.1 | |||
| AK014426.1 | 9 | 1369 | ||
| AK031671.1 | BAC27503.1 | |||
| BC023462.1 | AAH23462.1 | |||
| BC025109.1 | AAH25109.1 | |||
| BC058242.1 | AAH58242.1 |

Genomic Map