Assay Details
Target Gene Details
Entrez Gene ID: | 94184 |
Gene Name: | pyridoxal-dependent decarboxylase domain containing 1 |
Gene Aliases: |
2210010A19Rik, AA415817, Kiaa0251-hp |
Location: |
Chr.16:13833161-13903145 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdxdc1 | NM_001039533.2 | 1 | 91 | NP_001034622.1 |
| NM_001291017.1 | 1 | 91 | NP_001277946.1 | |
| NM_053181.3 | 1 | 91 | NP_444411.2 | |
| AK004611.1 | BAB23408.1 | |||
| AK054329.1 | 1 | 74 | BAC35732.1 | |
| AK163696.1 | BAE37462.1 | |||
| AK170661.1 | 1 | 78 | BAE41943.1 | |
| CJ181488.1 | 1 | 92 |

Genomic Map