Assay Details
Target Gene Details
Entrez Gene ID: | 239719 |
Gene Name: | MKL/myocardin-like 2 |
Gene Aliases: |
ENSMUSG00000075401, Gt4-1, MRTF-B, Mrtfb, mKIAA1243 |
Location: |
Chr.16:13256017-13417529 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mkl2 | NM_001122667.2 | NP_001116139.1 | ||
| NM_153588.3 | NP_705816.2 | |||
| AF532598.1 | AAN33042.1 | |||
| AK220377.1 | BAD90435.1 | |||
| BC131644.1 | ||||
| BC131645.1 |
More Information
Set Membership: |
|

Genomic Map