Assay Details
Target Gene Details
Entrez Gene ID: | 107589 |
Gene Name: | myosin, light polypeptide kinase |
Gene Aliases: |
9530072E15Rik, A930019C19Rik, AW489456, KRP, MLCK108, MLCK210, Mlck, nmMlck, smMLCK |
Location: |
Chr.16:34745199-35002436 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mylk | NM_139300.3 | 25 | 4705 | NP_647461.3 |
AK078556.1 | ||||
AK162008.1 | 7 | 1068 | BAE36678.1 | |
AY237726.1 | 25 | 4734 | AAO85807.1 | |
AY237727.2 | 10 | 2258 | AAO85808.1 | |
BC045197.1 | 10 | 2188 | AAH45197.1 | |
BC058610.1 | 25 | 4717 | AAH58610.2 | |
BC145279.1 | 25 | 4692 | ||
BC150749.1 | 25 | 4692 |
