Assay Details
Target Gene Details
Entrez Gene ID: | 208177 |
Gene Name: | pleckstrin homology-like domain, family B, member 2 |
Gene Aliases: |
AV253284, C820004H04Rik, LL5b, LL5beta |
Location: |
Chr.16:45746230-45953701 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phldb2 | NM_001252442.1 | 12 | 3037 | NP_001239371.1 |
| NM_153412.4 | 11 | 2878 | NP_700461.2 | |
| AF506821.1 | 11 | 2879 | AAM33635.1 | |
| AK035655.1 | 6 | 641 | BAC29141.1 | |
| AK050493.1 | 12 | 3045 | ||
| BC050915.1 | 12 | 2893 | AAH50915.1 | |
| BC060683.1 | 12 | 2893 | AAH60683.1 |

Genomic Map