Assay Details
Target Gene Details
Entrez Gene ID: | 67768 |
Gene Name: | N-6 adenine-specific DNA methyltransferase 1 (putative) |
Gene Aliases: |
5830445C04Rik, Hemk2, Pred28 |
Location: |
Chr.16:87354185-87368649 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| N6amt1 | NM_001159331.1 | NP_001152803.1 | ||
| NM_026366.2 | NP_080642.1 | |||
| AK012019.1 | ||||
| AK145617.1 | BAE26543.1 | |||
| AK160298.1 | ||||
| AY456393.1 | AAR19227.1 | |||
| AY536887.1 | AAS45233.1 | |||
| AY583759.1 | AAS94315.1 | |||
| BC098330.1 | AAH98330.1 | |||
| BC116393.1 | ||||
| BC116394.1 |
More Information
Set Membership: |
|

Genomic Map