Assay Details
Target Gene Details
Entrez Gene ID: | 171286 |
Gene Name: | solute carrier family 12 (potassium/chloride transporters), member 8 |
Gene Aliases: |
CCC9, E330020C02Rik |
Location: |
Chr.16:33517333-33664135 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc12a8 | NM_001083902.1 | NP_001077371.1 | ||
| NM_134251.2 | NP_599012.2 | |||
| AF319951.1 | AAL37178.1 | |||
| AK087781.1 | ||||
| AK165080.1 | BAE38029.1 | |||
| BC030926.1 | AAH30926.1 |
More Information
Set Membership: |
|

Genomic Map