Assay Details
Target Gene Details
Entrez Gene ID: | 27426 |
Gene Name: | N-acetylglucosamine-1-phosphodiester alpha-N-acetylglucosaminidase |
Gene Aliases: |
AI596180, UCE |
Location: |
Chr.16:5195280-5204040 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nagpa | NM_013796.3 | 5 | 893 | NP_038824.2 |
NR_027804.1 | 4 | 990 | ||
AK032158.1 | 5 | 894 | BAC27731.1 | |
AK087468.1 | 5 | 893 | ||
AK138035.1 | 5 | 886 | BAE23539.1 | |
AK155450.1 | 5 | 872 | BAE33269.1 | |
AK159629.1 | 5 | 872 | BAE35243.1 | |
AK162633.1 | 5 | 875 | ||
AK168830.1 | 4 | 978 | ||
AK169539.1 | 1 | 4316 | ||
AK170219.1 | 5 | 875 | BAE41643.1 | |
BC039790.1 | 5 | 885 | AAH39790.1 |
