Assay Details
Target Gene Details
Entrez Gene ID: | 67374 |
Gene Name: | junction adhesion molecule 2 |
Gene Aliases: |
1110002N23Rik, 2410030G21Rik, 2410167M24Rik, JAM-2, JAM-B, Jcam2, VE-JAM |
Location: |
Chr.16:84774123-84826380 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Jam2 | NM_023844.5 | 7 | 1107 | NP_076333.3 |
AF255911.1 | 7 | 830 | AAF81224.1 | |
AJ291757.1 | 7 | 1043 | CAC20699.1 | |
AK010616.1 | 7 | 949 | BAB27064.1 | |
AK013914.1 | 7 | 948 | BAB29053.1 | |
AK028757.1 | 7 | 798 | BAC26102.1 | |
AK078128.1 | 7 | 833 | BAC37139.1 | |
AK153636.1 | 7 | 798 | ||
BC028778.1 | 7 | 1107 | AAH28778.1 |
