Assay Details
Target Gene Details
Entrez Gene ID: | 13840 |
Gene Name: | Eph receptor A6 |
Gene Aliases: |
Ehk2, Hek12, m-ehk2 |
Location: |
Chr.16:59653061-60606484 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Epha6 |
