Assay Details
Target Gene Details
Entrez Gene ID: | 27281 |
Gene Name: | HRAS-like suppressor |
Gene Aliases: |
2810012B06Rik, A-C1, HRSL1, Hrasrs |
Location: |
Chr.16:29209695-29230531 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hrasls | NM_013751.5 | NP_038779.2 | ||
AK039471.1 | BAC30361.1 |
