Assay Details
Target Gene Details
Entrez Gene ID: | 27281 |
Gene Name: | HRAS-like suppressor |
Gene Aliases: |
2810012B06Rik, A-C1, HRSL1, Hrasrs |
Location: |
Chr.16:29209695-29230531 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hrasls | NM_013751.5 | 2 | 179 | NP_038779.2 |
| AB510982.1 | 1 | 64 | ||
| AF163095.1 | 1 | 64 | AAF13304.1 | |
| AK004528.1 | 2 | 213 | BAB23348.1 | |
| AK039471.1 | 2 | 180 | BAC30361.1 | |
| BC048482.1 | 2 | 183 | AAH48482.1 |

Genomic Map