Assay Details
Target Gene Details
Entrez Gene ID: | 17968 |
Gene Name: | neural cell adhesion molecule 2 |
Gene Aliases: |
Ncam-2, Ocam, RNCAM |
Location: |
Chr.16:81200697-81624290 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ncam2 | NM_001113208.1 | 9 | 1340 | NP_001106679.1 |
NM_010954.4 | 9 | 1340 | NP_035084.1 | |
AF001286.1 | 9 | 1266 | AAB69124.1 | |
AF001287.1 | 9 | 1266 | AAB69125.1 | |
AF016619.1 | 9 | 1222 | AAC53375.1 | |
BC119027.1 | 9 | 1198 | ||
BC119029.1 | 9 | 1198 |
