Assay Details
Target Gene Details
Entrez Gene ID: | 110920 |
Gene Name: | heat shock protein 70 family, member 13 |
Gene Aliases: |
1600002I10Rik, AV006182, B230217N24Rik, Stch |
Location: |
Chr.16:75755190-75767277 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspa13 | NM_030201.3 | NP_084477.1 | ||
| NR_027492.1 | ||||
| AK030553.1 | ||||
| AK034643.1 | BAC28781.1 | |||
| AK049011.1 | 3 | 2286 | ||
| AK167839.1 | BAE39860.1 | |||
| BC085181.1 | AAH85181.1 |

Genomic Map