Assay Details
Target Gene Details
Entrez Gene ID: | 50528 |
Gene Name: | transmembrane protease, serine 2 |
Gene Aliases: |
D16Ertd61e |
Location: |
Chr.16:97564682-97659538 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmprss2 | NM_015775.2 | NP_056590.2 | ||
| AF113596.1 | AAF21308.1 | |||
| AF199362.1 | AAF97867.1 | |||
| AF243500.1 | AAF64186.1 | |||
| AK141350.1 | ||||
| AK146047.1 | BAE26858.1 | |||
| AK161173.1 | BAE36225.1 | |||
| BC038393.1 | AAH38393.1 | |||
| BC054348.1 | AAH54348.1 |
More Information
Set Membership: |
|

Genomic Map