Assay Details
Target Gene Details
Entrez Gene ID: | 17174 |
Gene Name: | mannan-binding lectin serine peptidase 1 |
Gene Aliases: |
AW048060, CCPII, Crarf, Masp1/3 |
Location: |
Chr.16:23451785-23521073 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Masp1 | NM_008555.2 | NP_032581.2 | ||
| AB049755.1 | BAB69688.1 | |||
| AK031598.1 | BAC27469.1 | |||
| AK032944.1 | ||||
| AK043055.1 | ||||
| BC131637.1 | ||||
| BC131638.1 |
More Information
Set Membership: |
|

Genomic Map