Assay Details
Target Gene Details
Entrez Gene ID: | 224139 |
Gene Name: | golgi autoantigen, golgin subfamily b, macrogolgin 1 |
Gene Aliases: |
4930428L02Rik, 628101, 6330407A06Rik, AU042952, C130074L01Rik, F730017E11Rik, Gm6840, mKIAA4151 |
Location: |
Chr.16:36875173-36933085 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Golgb1 | NM_030035.1 | 20 | 9584 | NP_084311.1 |
AK081762.1 | ||||
AK085677.1 | ||||
AK220188.1 | 8 | 5439 | BAD90373.1 | |
BC040461.1 | 8 | 2904 | AAH40461.1 | |
BC042759.1 | 7 | 919 | AAH42759.1 | |
BC060254.1 | 7 | 2008 | AAH60254.1 | |
BC145221.1 | 18 | 2526 | ||
BC150731.1 | 20 | 9561 |
