Assay Details
Target Gene Details
Entrez Gene ID: | 68306 |
Gene Name: | RIKEN cDNA 4930565N06 gene |
Gene Aliases: |
4930428L02Rik |
Location: |
Chr.16:36880519-36897189 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930565N06Rik | NR_040476.1 | |||
| AK015360.1 | ||||
| AK019787.1 |
Target Gene Details
Entrez Gene ID: | 224139 |
Gene Name: | golgi autoantigen, golgin subfamily b, macrogolgin 1 |
Gene Aliases: |
4930428L02Rik, 628101, 6330407A06Rik, AU042952, C130074L01Rik, F730017E11Rik, Gm6840, mKIAA4151 |
Location: |
Chr.16:36875173-36933085 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Golgb1 | NM_030035.1 | NP_084311.1 | ||
| BC145221.1 | ||||
| BC150731.1 |
More Information
Set Membership: |
|

Genomic Map