Assay Details
Target Gene Details
Entrez Gene ID: | 433022 |
Gene Name: | phosphatidylinositol-specific phospholipase C, X domain containing 2 |
Gene Aliases: |
AW494639, EG433022 |
Location: |
Chr.16:45959261-46010413 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plcxd2 | NM_001134480.1 | 3 | 1257 | NP_001127952.1 |
BC151059.1 | 3 | 859 | ||
BC151063.1 | 3 | 859 |
