Assay Details
Target Gene Details
Entrez Gene ID: | 77626 |
Gene Name: | sphingomyelin phosphodiesterase 4 |
Gene Aliases: |
4122402O22Rik, mKIAA1418 |
Location: |
Chr.16:17619354-17644830 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smpd4 | NM_001164609.1 | NP_001158081.1 | ||
| NM_001164610.1 | NP_001158082.1 | |||
| NM_001164611.1 | NP_001158083.1 | |||
| NM_029945.3 | NP_084221.2 | |||
| AK019466.2 | BAB31737.1 | |||
| AK031029.1 | BAC27221.1 | |||
| AK031685.1 | BAC27512.1 | |||
| AK035892.1 | BAC29229.1 | |||
| AK036055.1 | ||||
| AK036380.1 | ||||
| AK129354.1 | BAC98164.1 | |||
| AK138720.1 | 16 | 2025 | ||
| AK164234.1 | BAE37694.1 | |||
| AK164456.1 | BAE37796.1 | |||
| AK182255.1 | ||||
| BC026767.1 | AAH26767.1 |
More Information
Set Membership: |
|

Genomic Map