Assay Details
Target Gene Details
Entrez Gene ID: | 545156 |
Gene Name: | kalirin, RhoGEF kinase |
Gene Aliases: |
2210407G14Rik, AV235988, DUET, E530005C20Rik, Gm539, Hapip, TRAD |
Location: |
Chr.16:33969073-34573291 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 54 - Exon 55 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kalrn | NM_177357.3 | NP_796331.2 |
More Information
Set Membership: |
|

Genomic Map