Assay Details
Target Gene Details
Entrez Gene ID: | 67474 |
Gene Name: | synaptosomal-associated protein 29 |
Gene Aliases: |
1300018G05Rik, AI891940, AU020222, BB131856, Gs32 |
Location: |
Chr.16:17406000-17430826 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snap29 | NM_023348.4 | 2 | 408 | NP_075837.3 |
| AK005042.1 | 2 | 358 | BAB23769.1 | |
| AK007065.1 | 2 | 401 | BAB24850.1 | |
| AK039203.1 | 2 | 377 | BAC30277.1 | |
| AK147845.1 | 2 | 316 | BAE28177.1 | |
| AY008722.1 | 2 | 405 | AAG32076.1 | |
| BC030066.1 | 2 | 357 | AAH30066.1 |
More Information
Set Membership: |
|

Genomic Map