Assay Details
Target Gene Details
Entrez Gene ID: | 74478 |
Gene Name: | sorting nexin 29 |
Gene Aliases: |
4933437K13Rik, ENSMUSG00000079738, Gm11170, Gm1737, Gm930, Rundc2a |
Location: |
Chr.16:11321256-11755931 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snx29 | NM_001290148.1 | 6 | 554 | NP_001277077.1 |
| NM_028964.4 | 13 | 1603 | NP_083240.2 | |
| AF399755.1 | 6 | 509 | AAK94018.1 | |
| AK017102.1 | 6 | 555 | BAB30596.1 | |
| AK086865.1 | 1 | 136 | ||
| AK162238.1 | 5 | 432 | BAE36809.1 | |
| BB623950.1 | 5 | 432 | ||
| BC034114.1 | 3 | 203 | AAH34114.1 |
More Information
Set Membership: |
|

Genomic Map