Assay Details
Target Gene Details
Entrez Gene ID: | 22195 |
Gene Name: | ubiquitin-conjugating enzyme E2L 3 |
Gene Aliases: |
C79827, UbcM4, Ubce7 |
Location: |
Chr.16:17152015-17201498 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2l3 | NM_009456.2 | NP_033482.1 | ||
| AJ130961.1 | CAA10265.1 | |||
| AK007265.1 | BAB24925.1 | |||
| AK152522.1 | BAE31282.1 | |||
| AK165620.1 | BAE38300.1 | |||
| AK167847.1 | BAE39867.1 | |||
| BC093503.1 | AAH93503.1 | |||
| BC106149.1 | AAI06150.1 | |||
| X97042.1 | CAA65755.1 |
More Information
Set Membership: |
|

Genomic Map