Assay Details
Target Gene Details
Entrez Gene ID: | 14805 |
Gene Name: | glutamate receptor, ionotropic, kainate 1 |
Gene Aliases: |
A830007B11Rik, D16Ium24, D16Ium24e, GluK1, GluK5, Glur-5, Glur5, Glurbeta1 |
Location: |
Chr.16:87895897-88290602 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grik1 | NM_010348.3 | NP_034478.1 | ||
| NM_146072.4 | NP_666184.2 | |||
| AK042635.1 | BAC31315.1 | |||
| AK043542.1 | BAC31575.1 | |||
| AK043737.1 | BAC31636.1 | |||
| AK044250.1 | ||||
| AK044420.1 | ||||
| AK046122.1 | BAC32609.1 | |||
| AK048634.1 | BAC33402.1 | |||
| AK133667.1 | BAE21774.1 | |||
| AK200858.1 | ||||
| BC031822.1 | AAH31822.1 | |||
| BC049275.1 | AAH49275.1 | |||
| X66118.1 |
More Information
Set Membership: |
|

Genomic Map