Assay Details
Target Gene Details
Entrez Gene ID: | 210530 |
Gene Name: | prolyl 3-hydroxylase 2 |
Gene Aliases: |
4832416N06, AW553532, Leprel1, Mlat4 |
Location: |
Chr.16:25960323-26105784 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P3h2 | NM_173379.2 | NP_775555.1 | ||
| AJ430350.1 | CAD23038.1 |
More Information
Set Membership: |
|

Genomic Map