Assay Details
Target Gene Details
Entrez Gene ID: | 239759 |
Gene Name: | lipase, member H |
Gene Aliases: |
C130037N08Rik, D16Wsu119e, H06, Lpdlr, P3, PLA1B, lpd1, lpd2, mPA-PLA1 |
Location: |
Chr.16:21953817-21995542 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Liph | NM_001083894.1 | NP_001077363.1 | ||
| NM_001289581.1 | NP_001276510.1 | |||
| NM_001289582.1 | NP_001276511.1 | |||
| NM_153404.3 | NP_700453.1 | |||
| AK035498.1 | ||||
| AK048150.1 | BAC33259.1 | |||
| AK134352.1 | BAE22108.1 | |||
| AK136503.1 | BAE23014.1 | |||
| AK137278.1 | ||||
| AK161067.1 | BAE36182.1 | |||
| AK162492.1 | BAE36944.1 | |||
| AY093499.1 | AAM18804.1 | |||
| BC037489.1 | AAH37489.1 |
More Information
Set Membership: |
|

Genomic Map