Assay Details
Target Gene Details
Entrez Gene ID: | 21976 |
Gene Name: | topoisomerase (DNA) III beta |
Gene Aliases: |
- |
Location: |
Chr.16:16870694-16892986 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Top3b | NM_011624.2 | NP_035754.1 | ||
| AB013603.1 | BAA34227.1 | |||
| AB045324.1 | BAA97984.1 | |||
| AK144709.1 | BAE26026.1 | |||
| AK146475.1 | BAE27200.1 | |||
| AK163716.1 | ||||
| AK163796.1 | ||||
| BC031723.1 | AAH31723.1 |
More Information
Set Membership: |
|

Genomic Map