Assay Details
Target Gene Details
Entrez Gene ID: | 381072 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 17 |
Gene Aliases: |
- |
Location: |
Chr.17:24264258-24351897 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abca17 | NM_001031621.2 | NP_001026792.2 | ||
| AB112584.1 | BAD97416.1 |
More Information
Set Membership: |
|

Genomic Map