Assay Details
Target Gene Details
Entrez Gene ID: | 56298 |
Gene Name: | atlastin GTPase 2 |
Gene Aliases: |
2010110I21Rik, AA407293, AV334690, Aip-2, Arl6ip2 |
Location: |
Chr.17:79848352-79896123 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atl2 | NM_001286647.1 | 1 | 116 | NP_001273576.1 |
| NM_019717.3 | 1 | 116 | NP_062691.3 | |
| NM_178050.4 | 1 | 116 | NP_835151.2 | |
| AK008381.1 | 1 | 117 | BAB25636.1 | |
| AK038697.1 | 1 | 44 | BAC30103.1 | |
| AK043907.1 | 1 | 105 | ||
| AK135988.1 | BAE22763.1 | |||
| BC060501.1 | AAH60501.1 | |||
| BY244533.1 | 1 | 114 |
More Information
Set Membership: |
|

Genomic Map