Assay Details
Target Gene Details
Entrez Gene ID: | 74123 |
Gene Name: | forkhead box P4 |
Gene Aliases: |
1200010K03Rik, 2310007G05Rik, mFKHLA |
Location: |
Chr.17:47867133-47924645 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Foxp4 | NM_001110824.1 | 16 | 2223 | NP_001104294.1 |
NM_001110825.1 | 16 | 2187 | NP_001104295.1 | |
NM_028767.2 | 16 | 2184 | NP_083043.2 | |
AB052766.1 | 16 | 2018 | BAC53799.1 | |
AK004693.1 | 16 | 1995 | BAB23479.1 | |
AK143194.1 | 16 | 2035 | ||
AY135029.1 | 16 | 2011 | AAN08624.1 | |
BC043702.1 | 16 | 1961 | AAH43702.1 | |
BC052407.1 | 16 | 2026 | AAH52407.1 | |
BC057110.1 | 16 | 2184 | AAH57110.1 |
