Assay Details
Target Gene Details
Entrez Gene ID: | 12575 |
Gene Name: | cyclin-dependent kinase inhibitor 1A (P21) |
Gene Aliases: |
CAP20, CDKI, CIP1, Cdkn1, P21, SDI1, Waf1, mda6, p21Cip1, p21WAF |
Location: |
Chr.17:29090986-29100722 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdkn1a | NM_001111099.1 | NP_001104569.1 | ||
| BY206886.1 |
More Information
Set Membership: |
|

Genomic Map