Assay Details
Target Gene Details
Entrez Gene ID: | 22247 |
Gene Name: | uridine monophosphate synthetase |
Gene Aliases: |
1700095D23Rik, AA408257, AL033308, BB164745 |
Location: |
Chr.16:33954784-33967302 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Umps | NM_009471.2 | NP_033497.1 | ||
| AK035821.1 | BAC29199.1 | |||
| AK035997.1 | ||||
| AK090102.1 | ||||
| AK159808.1 | BAE35388.1 | |||
| AK159955.1 | BAE35511.1 | |||
| AK183780.1 | ||||
| BC003887.1 | AAH03887.1 | |||
| BC095981.1 | AAH95981.1 | |||
| M29395.1 | AAA39859.1 |
More Information
Set Membership: |
|

Genomic Map