Assay Details
Target Gene Details
Entrez Gene ID: | 19106 |
Gene Name: | eukaryotic translation initiation factor 2-alpha kinase 2 |
Gene Aliases: |
2310047A08Rik, 4732414G15Rik, AI467567, AI747578, Pkr, Prkr, Tik |
Location: |
Chr.17:78852550-78882572 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif2ak2 | NM_011163.4 | 5 | 631 | NP_035293.1 |
| AK028602.1 | 5 | 631 | BAC26027.1 | |
| BC016422.1 | 5 | 594 | AAH16422.1 | |
| M65029.1 | 5 | 583 | AAA40150.1 | |
| M93567.1 | 5 | 573 | AAA39885.1 |
More Information
Set Membership: |
|

Genomic Map