Assay Details
Target Gene Details
Entrez Gene ID: | 381122 |
Gene Name: | calpain 13 |
Gene Aliases: |
CANP 13, Gm943 |
Location: |
Chr.17:73306456-73399306 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Capn13 | NM_001033444.2 | NP_001028616.1 | ||
| AK136569.1 | BAE23051.1 | |||
| BC147510.1 | ||||
| BC147512.1 |
More Information
Set Membership: |
|

Genomic Map