Assay Details
Target Gene Details
Entrez Gene ID: | 68597 |
Gene Name: | coiled-coil domain containing 167 |
Gene Aliases: |
1110021J02Rik |
Location: |
Chr.17:29695977-29717017 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc167 | NM_001163741.2 | 5 | 3433 | NP_001157213.1 |
Target Gene Details
Entrez Gene ID: | 74157 |
Gene Name: | cap methyltransferase 1 |
Gene Aliases: |
1300018I05Rik, AI451264, Ftsjd2, MTr1, mKIAA0082 |
Location: |
Chr.17:29660601-29703359 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cmtr1 | NM_028791.5 | 17 | 1931 | NP_083067.1 |
| AK005044.2 | 17 | 1838 | BAB23771.1 | |
| AK042060.1 | 17 | 1877 | BAC31146.1 | |
| AK043733.1 | 17 | 1868 | BAC31634.1 | |
| AK049190.1 | 17 | 1915 | BAC33600.1 | |
| AK080379.1 | 17 | 1932 | BAC37899.1 | |
| AK088624.1 | 17 | 1866 | ||
| AK150392.1 | 17 | 1857 | BAE29520.1 | |
| AK151194.1 | 17 | 1857 | BAE30191.1 | |
| AK152276.1 | 17 | 1857 | BAE31091.1 | |
| AK152452.1 | 17 | 1838 | BAE31230.1 | |
| AK152799.1 | 17 | 1857 | BAE31504.1 | |
| AK153021.1 | 17 | 1857 | BAE31655.1 | |
| AK153066.1 | 17 | 1857 | BAE31692.1 | |
| AK154773.1 | 17 | 1903 | BAE32821.1 | |
| AK172883.1 | 14 | 1413 | BAD32161.1 | |
| BC024691.1 | 17 | 1853 | AAH24691.1 |

Genomic Map