Assay Details
Target Gene Details
Entrez Gene ID: | 73242 |
Gene Name: | alpha tubulin acetyltransferase 1 |
Gene Aliases: |
0610011P08Rik, 2610008K08Rik, 2610110G12Rik, 3110080J08Rik, Mec17, TAT |
Location: |
Chr.17:35897595-35910093 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atat1 | NM_001142744.1 | 10 | 838 | NP_001136216.1 |
| NM_001142745.1 | 10 | 838 | NP_001136217.1 | |
| NM_028476.4 | 9 | 769 | NP_082752.3 | |
| AK011350.1 | 9 | 743 | ||
| AK011750.2 | 10 | 814 | ||
| AK011838.1 | 10 | 827 | ||
| AK014265.1 | 10 | 812 | ||
| AK028297.1 | 10 | 828 | BAC25866.1 | |
| AK034725.1 | 9 | 747 | BAC28808.1 | |
| AK042804.1 | 4 | 264 | ||
| AK049682.1 | 9 | 719 | ||
| AK133694.1 | 10 | 815 | BAE21786.1 | |
| BC028847.1 | 10 | 817 | AAH28847.1 | |
| BC030669.1 | 9 | 747 | AAH30669.1 |

Genomic Map