Assay Details
Target Gene Details
Entrez Gene ID: | 53817 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B |
Gene Aliases: |
0610030D10Rik, AI428441, Bat-1, Bat1, Bat1a, D17H6S81E, D17H6S81E-1, D6S81Eh |
Location: |
Chr.17:35241746-35253707 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx39b | NM_001252457.1 | NP_001239386.1 | ||
| NM_019693.3 | NP_062667.1 | |||
| AF118128.1 | AAD13115.1 | |||
| AK051034.1 | BAC34505.1 | |||
| AK088867.1 | BAC40624.1 | |||
| AY255786.1 | AAP91685.1 | |||
| AY255787.1 | AAP91686.1 | |||
| BC011067.1 | AAH11067.1 | |||
| BC024859.1 | AAH24859.1 |
Target Gene Details
Entrez Gene ID: | 102465886 |
Gene Name: | microRNA 8094 |
Gene Aliases: |
Gm27513, mmu-mir-8094 |
Location: |
Chr.17:35251289-35251405 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir8094 | NR_106169.1 |

Genomic Map