Assay Details
Target Gene Details
Entrez Gene ID: | 68597 |
Gene Name: | coiled-coil domain containing 167 |
Gene Aliases: |
1110021J02Rik |
Location: |
Chr.17:29695977-29717017 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc167 | NM_001163741.2 | 5 | 2585 | NP_001157213.1 |
AK085526.1 | 5 | 2666 | BAC39463.1 |
Target Gene Details
Entrez Gene ID: | 74157 |
Gene Name: | cap methyltransferase 1 |
Gene Aliases: |
1300018I05Rik, AI451264, Ftsjd2, MTr1, mKIAA0082 |
Location: |
Chr.17:29660601-29703359 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cmtr1 | NM_028791.5 | 18 | 2046 | NP_083067.1 |
AK005044.2 | 18 | 1953 | BAB23771.1 | |
AK042060.1 | 18 | 1992 | BAC31146.1 | |
AK043733.1 | 18 | 1983 | BAC31634.1 | |
AK049190.1 | 18 | 2030 | BAC33600.1 | |
AK080379.1 | 18 | 2047 | BAC37899.1 | |
AK088624.1 | 18 | 1981 | ||
AK150392.1 | 18 | 1972 | BAE29520.1 | |
AK151194.1 | 18 | 1972 | BAE30191.1 | |
AK152276.1 | 18 | 1972 | BAE31091.1 | |
AK152452.1 | 18 | 1953 | BAE31230.1 | |
AK152799.1 | 18 | 1972 | BAE31504.1 | |
AK153021.1 | 18 | 1972 | BAE31655.1 | |
AK153066.1 | 18 | 1972 | BAE31692.1 | |
AK154773.1 | 18 | 2018 | BAE32821.1 | |
AK172883.1 | 15 | 1528 | BAD32161.1 | |
BC024691.1 | 18 | 1968 | AAH24691.1 |
