Assay Details
Target Gene Details
Entrez Gene ID: | 74157 |
Gene Name: | cap methyltransferase 1 |
Gene Aliases: |
1300018I05Rik, AI451264, Ftsjd2, MTr1, mKIAA0082 |
Location: |
Chr.17:29660601-29703359 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cmtr1 | NM_028791.5 | NP_083067.1 | ||
| AK005044.2 | BAB23771.1 | |||
| AK042060.1 | BAC31146.1 | |||
| AK043733.1 | BAC31634.1 | |||
| AK049190.1 | BAC33600.1 | |||
| AK080379.1 | BAC37899.1 | |||
| AK088624.1 | ||||
| AK150392.1 | BAE29520.1 | |||
| AK151194.1 | BAE30191.1 | |||
| AK152276.1 | BAE31091.1 | |||
| AK152452.1 | BAE31230.1 | |||
| AK152799.1 | BAE31504.1 | |||
| AK153021.1 | BAE31655.1 | |||
| AK153066.1 | BAE31692.1 | |||
| AK154773.1 | BAE32821.1 | |||
| AK172883.1 | BAD32161.1 | |||
| BC024691.1 | AAH24691.1 |

Genomic Map