Assay Details
Target Gene Details
Entrez Gene ID: | 14962 |
Gene Name: | complement factor B |
Gene Aliases: |
AI195813, AI255840, B, Bf, C2, Fb, H2-Bf |
Location: |
Chr.17:34856374-34862514 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfb | NM_001142706.1 | 8 | 1504 | NP_001136178.1 |
| NM_008198.2 | 8 | 1504 | NP_032224.2 | |
| AK098094.1 | 8 | 1164 | ||
| AK149531.1 | 8 | 1137 | BAE28943.1 | |
| AK149573.1 | 8 | 1163 | ||
| AK151220.1 | 2 | 123 | BAE30214.1 | |
| AK151246.1 | 8 | 1238 | BAE30236.1 | |
| AK151618.1 | 2 | 123 | BAE30555.1 | |
| AK151974.1 | 2 | 123 | BAE30842.1 | |
| BC005451.1 | 8 | 1138 | AAH05451.1 | |
| K01496.1 | 3 | 244 | AAA39549.1 | |
| K01497.1 | 2 | 103 | AAA39550.1 | |
| M57890.1 | 8 | 1207 | AAA63293.1 |

Genomic Map