Assay Details
Target Gene Details
Entrez Gene ID: | 14158 |
Gene Name: | fer (fms/fps related) protein kinase |
Gene Aliases: |
AV082135, C330004K01Rik, Fert, Fert2 |
Location: |
Chr.17:63829950-64146519 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fer | NM_001037997.3 | 11 | 1602 | NP_001033086.2 |
NM_001286415.1 | 10 | 1430 | NP_001273344.1 | |
NM_008000.2 | 6 | 884 | NP_032026.2 | |
AF286537.1 | AAG40730.1 | |||
AK082799.1 | 8 | 1095 | BAC38626.1 | |
AK157828.1 | 11 | 1566 | BAE34213.1 | |
BC051249.1 | 3 | 346 | AAH51249.1 | |
BC058100.1 | 10 | 1430 | AAH58100.1 | |
M32054.1 | 6 | 884 | AAA37617.1 | |
U76762.1 | 10 | 1535 | AAB18988.1 |
